Assay Details
| Gene Symbol: Cenpu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027973.3 | NP_082249.1 | 1 - 2 | 56 | 101 | |
| GenBank mRNA | AK006479.1 | - | 1 - 2 | 65 | 101 | |
| AK050386.1 | - | 1 - 2 | 57 | 101 | ||
| AK081703.1 | - | 1 - 2 | 44 | 101 | ||
| BC117784.2 | - | 1 - 2 | 57 | 40092112 | 101 | |
| BC117785.2 | - | 1 - 2 | 57 | 40092113 | 101 | |
More Information
Set Membership: |
|

Cenpu
Genomic Map