Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800003807.mm.1 |
NM_027973.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003807.mm.1 |
NM_027973.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003807.mm.1 |
TC0800000539.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800003809.mm.1 |
NM_027973.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003809.mm.1 |
NM_027973.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003809.mm.1 |
TC0800000539.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800003805.mm.1 |
NM_027973.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003805.mm.1 |
NM_027973.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003805.mm.1 |
TC0800000539.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800003804.mm.1 |
NM_027973.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003804.mm.1 |
NM_027973.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003804.mm.1 |
TC0800000539.mm.1 |
MTA TC |