Assay Details
Gene Symbol: Zfat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145888.1 | NP_001139360.1 | 11 - 12 | 3132 | 113 | |
NM_198644.2 | NP_941046.1 | 11 - 12 | 3111 | 113 | ||
GenBank mRNA | BC053930.1 | - | 11 - 12 | 3102 | 6389756 | 113 |
EU221277.1 | - | 11 - 12 | 2962 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
