Assay Details
| Gene Symbol: Gpsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029522.2 | NP_083798.2 | 4 - 5 | 914 | 125 | |
| GenBank mRNA | AK040996.1 | - | 4 - 5 | 803 | 125 | |
| AK143552.1 | - | 4 - 5 | 915 | 125 | ||
| AY081187.1 | - | 4 - 5 | 758 | 125 | ||
| BC021308.1 | - | 3 - 4 | 469 | 5007832 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpsm2
Genomic Map