Assay Details
| Gene Symbol: Tle4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF229633.1 | - | 10 - 11 | 796 | 107 | |
| AK122481.2 | - | 8 - 9 | 655 | 107 | ||
| AK134989.1 | - | 10 - 11 | 1430 | 107 | ||
| BC086631.1 | - | 11 - 12 | 1340 | 107 | ||
| U61363.1 | - | 2 - 3 | 199 | 107 | ||
More Information
Set Membership: |
|

Tle4
Genomic Map