Assay Details
Gene Symbol: Taf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009315.3 | NP_033341.1 | 6 - 7 | 648 | 125 | |
GenBank mRNA | AK153731.1 | - | 696 | 125 | ||
AK154867.1 | - | 6 - 7 | 682 | 125 | ||
BC058583.1 | - | 6 - 7 | 648 | 6514960 | 125 | |
D49439.1 | - | 5 - 6 | 581 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
