Assay Details
| Gene Symbol: Gtf2i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080746.2 | NP_001074215.1 | 30 - 31 | 3204 | 96 | |
| NM_001080747.2 | NP_001074216.1 | 29 - 30 | 3141 | 96 | ||
| NM_001080748.2 | NP_001074217.1 | 28 - 29 | 3084 | 96 | ||
| NM_001080749.2 | NP_001074218.1 | 28 - 29 | 3096 | 96 | ||
| NM_010365.4 | NP_034495.2 | 29 - 30 | 3147 | 96 | ||
| GenBank mRNA | AF017085.1 | - | 30 - 31 | 2848 | 96 | |
| AF043219.1 | - | 28 - 29 | 2768 | 96 | ||
| AF043220.1 | - | 27 - 28 | 2705 | 96 | ||
| AK013348.1 | - | 7 - 8 | 681 | 96 | ||
| AK145698.1 | - | 20 - 21 | 2032 | 96 | ||
| AK147201.1 | - | 28 - 29 | 3054 | 96 | ||
| AY030290.1 | - | 29 - 30 | 2834 | 96 | ||
| AY030291.1 | - | 29 - 30 | 2840 | 96 | ||
| AY030292.1 | - | 28 - 29 | 2777 | 96 | ||
| AY030293.1 | - | 30 - 31 | 2897 | 96 | ||
| BC047387.1 | - | 28 - 29 | 2743 | 96 | ||
| BC053044.1 | - | 30 - 31 | 3173 | 5704935 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2i
Genomic Map