Assay Details
| Gene Symbol: Lmtk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081109.1 | NP_001074578.1 | 2 - 3 | 466 | 133 | |
| GenBank mRNA | AB288872.1 | - | 2 - 3 | 294 | 133 | |
| AK158724.1 | - | 2 - 3 | 467 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lmtk2
Genomic Map