Assay Details
Gene Symbol: Lmtk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081109.1 | NP_001074578.1 | 3 - 4 | 608 | 59 | |
GenBank mRNA | AB288872.1 | - | 3 - 4 | 436 | 59 | |
AK158724.1 | - | 3 - 4 | 609 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
