Assay Details
| Gene Symbol: Pla2g4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004762.3 | NP_001004762.1 | 4 - 5 | 541 | 89 | |
| NM_001168504.1 | NP_001161976.1 | 4 - 5 | 557 | 89 | ||
| GenBank mRNA | AK136098.1 | - | 4 - 5 | 542 | 89 | |
| AK136143.1 | - | 4 - 5 | 544 | 89 | ||
| AK145339.1 | - | 4 - 5 | 542 | 89 | ||
| AK162196.1 | - | 4 - 5 | 544 | 89 | ||
| AY694793.1 | - | 4 - 5 | 528 | 89 | ||
| BC054740.1 | - | 2 - 3 | 254 | 89 | ||
| BC117808.1 | - | 4 - 5 | 557 | 40092567 | 89 | |
| BC117809.1 | - | 4 - 5 | 557 | 40092569 | 89 | |
| BC129974.1 | - | 4 - 5 | 557 | 40092571 | 89 | |
| BC138548.1 | - | 4 - 5 | 517 | 8861568 | 89 | |
| BC145767.1 | - | 4 - 5 | 517 | 40131070 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g4c
Genomic Map