Assay Details
| Gene Symbol: Glyat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145935.3 | NP_666047.1 | 3 - 4 | 502 | 137 | |
| GenBank mRNA | AK039262.1 | - | 3 - 4 | 503 | 137 | |
| AK143803.1 | - | 292 | 137 | |||
| AK143901.1 | - | 292 | 137 | |||
| BC010799.1 | - | 3 - 4 | 277 | 4234793 | 137 | |
| BC015294.1 | - | 314 | 4220471 | 137 | ||
| BC024434.1 | - | 2 - 3 | 192 | 4972912 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318856 | BC015294.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318856 | BC024434.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318856 | AK143803.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318856 | AK143901.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318856 | BC010799.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318856 | BC015294.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318856 | BC024434.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318856 | AK143803.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318856 | AK143901.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318856 | BC010799.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 23544 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717829 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 228594 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36964 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703265 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207266 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 369522 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597427 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955591 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330076 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538556 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138242 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 53749 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46129 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137364 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138963 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318856 | BC015294.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318856 | BC024434.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318856 | AK143803.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318856 | AK143901.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318856 | BC010799.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318856 | BC015294.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318856 | BC024434.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318856 | AK143803.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318856 | AK143901.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318856 | BC010799.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318856 | BC015294.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318856 | BC024434.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318856 | AK143803.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318856 | AK143901.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318856 | BC010799.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318856 | BC015294.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318856 | BC024434.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318856 | AK143803.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318856 | AK143901.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318856 | BC010799.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961677 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137364 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138963 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09108930 | - | - |
| Whole Mouse Genome G4122A | A_51_P318856 | BC015294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318856 | BC024434.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318856 | AK143803.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318856 | AK143901.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318856 | BC010799.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318856 | BC015294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318856 | BC024434.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318856 | AK143803.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318856 | AK143901.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318856 | BC010799.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P506984 | AK143901.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107089 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138510 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137364 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138963 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14463078 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14307747 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14463079 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14184576 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14363757 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14463072 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14463076 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14463080 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14307748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961712 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14418727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14313354 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14463080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14259833 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14402871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14276438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14265918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961710 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10461742 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461738 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461744 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461741 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461736 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461737 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461743 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461740 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424867_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451515_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424868_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10461735 | - | - |
| Mouse Genome 430 2.0 Array | 1451515_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1451515_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1424867_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1424867_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1424868_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424868_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107146_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107146_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900002960.mm.1 | NM_145935.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900002960.mm.1 | NM_145935.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900002960.mm.1 | TC1900000250.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900002962.mm.1 | NM_145935.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900002962.mm.1 | NM_145935.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900002962.mm.1 | TC1900000250.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900002964.mm.1 | TC1900000250.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900002965.mm.1 | TC1900000250.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900002958.mm.1 | NM_145935.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900002958.mm.1 | NM_145935.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900002958.mm.1 | TC1900000250.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430A Array | 1424867_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451515_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424868_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451515_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424868_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424867_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 520033 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6200239 | - | - |
| MouseWG-6 v2 Expression BeadChip | 103060152 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103060152 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6200239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006200239 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2697749 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1235058 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2697749 | - | - |
| mouse-6 v1.1 (Illumina) | scl00107146.2_299-S | - | - |
| mouse-6 v1.1 (Illumina) | scl53339.6_23-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103060152 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6200239 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00107146.2_299-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103060152 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6200239 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2697749 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2697749 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1235058 | - | - |

Glyat
Genomic Map