Assay Details
| Gene Symbol: Dpcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172639.2 | NP_766227.1 | 1 - 2 | 96 | 112 | |
| GenBank mRNA | AK077347.1 | - | 1 - 2 | 97 | 112 | |
| BC027559.1 | - | 1 - 2 | 108 | 1493971 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpcd
Genomic Map