Assay Details
| Gene Symbol: Dpcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172639.2 | NP_766227.1 | 5 - 6 | 537 | 61 | |
| GenBank mRNA | AK037566.1 | - | 2 - 3 | 185 | 61 | |
| AK077347.1 | - | 5 - 6 | 538 | 61 | ||
| BC027559.1 | - | 5 - 6 | 538 | 1493971 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P37498 | NM_172639.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P37498 | AK077347.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P37498 | BC027559.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P37498 | NM_172639.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P37498 | AK077347.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P37498 | BC027559.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 345174 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773587 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 24054 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 505810 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 929745 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 858724 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 961526 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 111395 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580493 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193900 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67281 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 17955 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 63506 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171736 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166620 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P37498 | NM_172639.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P37498 | AK077347.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P37498 | BC027559.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P37498 | NM_172639.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P37498 | AK077347.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P37498 | BC027559.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32139165 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03605383 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171736 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03605376 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166620 | - | - |
| Whole Mouse Genome G4122A | A_52_P37498 | NM_172639.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P37498 | AK077347.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P37498 | BC027559.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P37498 | NM_172639.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P37498 | AK077347.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P37498 | BC027559.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P369294 | NM_172639.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P369294 | AK077347.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118283 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166620 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171736 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14300077 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14168955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32139226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14481379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14246864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14235465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14481338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14150683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14481323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14144108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14330216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14481324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14330218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14218628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14354523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14246862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14481343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14208895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14481356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14380513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32139194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14481362 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10463510 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10463509 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10463507 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10463511 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10463508 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10463512 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10463506 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452603_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10463505 | - | - |
| Mouse Genome 430 2.0 Array | 1442674_at | - | - |
| Mouse Genome 430 2.0 Array | 1442674_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452603_at | - | - |
| Mouse Genome 430 2.0 Array | 1452603_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 226162_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 226162_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900005958.mm.1 | NM_172639.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900005958.mm.1 | NM_172639.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900005958.mm.1 | TC1900000675.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900005957.mm.1 | NM_172639.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900005957.mm.1 | NM_172639.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900005957.mm.1 | TC1900000675.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900005956.mm.1 | NM_172639.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900005956.mm.1 | NM_172639.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900005956.mm.1 | TC1900000675.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452603_PM_at | - | - |
| Mouse Expression Set 430 | 1452603_at | - | - |
| Murine Genome U74v2 Set | 104120_PM_at | - | - |
| Murine Genome U74v2 Set | 104120_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452603_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452603_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452603_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452603_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 576732 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2120452 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 780632 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2120452 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002120452 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000780632 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237049 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2705644 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237049 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2705644 | - | - |
| mouse-6 v1.1 (Illumina) | scl0226162.6_83-S | - | - |
| mouse-6 v1.1 (Illumina) | scl000481.1_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 780632 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2120452 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0226162.6_83-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000481.1_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2120452 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 780632 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2892480 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2892479 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1237049 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2892480 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2892479 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237049 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl000481.1_29-S | - | - |

Dpcd
Genomic Map