Assay Details
Gene Symbol: Lat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020044.3 | NP_064428.1 | 11 - 12 | 693 | 65 | |
NM_022964.4 | NP_075253.2 | 10 - 11 | 657 | 65 | ||
GenBank mRNA | AF257136.1 | - | 10 - 11 | 637 | 65 | |
AK134721.1 | - | 8 - 9 | 385 | 65 | ||
AK138953.1 | - | 11 - 12 | 652 | 65 | ||
AK143533.1 | - | 10 - 11 | 658 | 65 | ||
AK144261.1 | - | 11 - 12 | 632 | 65 | ||
AK162321.1 | - | 10 - 11 | 616 | 65 | ||
AY190024.1 | - | 11 - 12 | 658 | 65 | ||
BC005804.1 | - | 10 - 11 | 586 | 3154073 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
