Assay Details
Gene Symbol: Lat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020044.3 | NP_064428.1 | 12 - 13 | 803 | 80 | |
NM_022964.4 | NP_075253.2 | 11 - 12 | 767 | 80 | ||
GenBank mRNA | AF257136.1 | - | 11 - 12 | 747 | 80 | |
AK134721.1 | - | 9 - 10 | 495 | 80 | ||
AK138953.1 | - | 12 - 13 | 762 | 80 | ||
AK143533.1 | - | 11 - 12 | 768 | 80 | ||
AK144261.1 | - | 12 - 13 | 742 | 80 | ||
AK162321.1 | - | 11 - 12 | 726 | 80 | ||
AY190024.1 | - | 12 - 13 | 768 | 80 | ||
BC005804.1 | - | 11 - 12 | 696 | 3154073 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242930 | AK144261.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242930 | AK162321.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242930 | BC005804.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242930 | NM_022964.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242930 | AK134721.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242930 | AK144261.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242930 | AK162321.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242930 | BC005804.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242930 | NM_022964.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P242930 | AK134721.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917710 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 11495 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445359 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323418 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 128430 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 797759 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 793491 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 398154 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 82278 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 47282 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 748313 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 105258 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 5537 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160173 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242930 | AK144261.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242930 | AK162321.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242930 | BC005804.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242930 | NM_022964.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242930 | AK134721.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242930 | AK144261.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242930 | AK162321.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242930 | BC005804.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242930 | NM_022964.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P242930 | AK134721.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P242930 | AK144261.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P242930 | AK162321.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P242930 | BC005804.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P242930 | NM_022964.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P242930 | AK134721.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P242930 | AK144261.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P242930 | AK162321.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P242930 | BC005804.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P242930 | NM_022964.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P242930 | AK134721.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09573388 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03804106 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03804117 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09573403 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160173 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03804112 | - | - |
Whole Mouse Genome G4122A | A_51_P242930 | AK144261.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P242930 | AK162321.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P242930 | BC005804.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P242930 | NM_022964.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P242930 | AK134721.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P242930 | AK144261.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P242930 | AK162321.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P242930 | BC005804.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P242930 | NM_022964.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P242930 | AK134721.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P426049 | AK144261.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P426049 | AK162321.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P426049 | AY190024.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P426049 | BC005804.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P426049 | NM_022964.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120625 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160173 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03804117 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001133 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001118 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001135 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001132 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001134 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001147 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001155 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001125 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001130 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001144 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001150 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001139 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001156 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001142 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001137 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001153 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001136 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001123 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001128 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001145 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001121 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001120 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001152 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001138 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P33001141 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10534306 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534310 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534304 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534314 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534307 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534313 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534315 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534309 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534312 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534311 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534308 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534305 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449606_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1443039_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1426169_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1443040_PM_x_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10534303 | - | - |
Mouse Genome 430 2.0 Array | 1443040_x_at | - | - |
Mouse Genome 430 2.0 Array | 1426169_a_at | - | - |
Mouse Genome 430 2.0 Array | 1449606_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1443040_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1443039_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1443039_at | - | - |
Mouse Genome 430 2.0 Array | 1449606_at | - | - |
Mouse Genome 430 2.0 Array | 1426169_PM_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56743_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56743_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500030430.mm.1 | TC0500003376.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500030430.mm.1 | NM_020044.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500030430.mm.1 | NM_022964.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500030429.mm.1 | TC0500003376.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500030429.mm.1 | NM_020044.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500030429.mm.1 | NM_022964.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500030427.mm.1 | TC0500003376.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500030427.mm.1 | NM_020044.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500030427.mm.1 | NM_022964.4 | Refseq |
Mouse Expression Set 430 | 1443040_x_at | - | - |
Mouse Expression Set 430 | 1449606_PM_at | - | - |
Mouse Expression Set 430 | 1443039_at | - | - |
Mouse Expression Set 430 | 1443039_PM_at | - | - |
Mouse Expression Set 430 | 1449606_at | - | - |
Mouse Expression Set 430 | 1443040_PM_x_at | - | - |
Murine Genome U74v2 Set | 161899_PM_f_at | - | - |
Murine Genome U74v2 Set | 161899_f_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426169_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426169_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1449606_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443040_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1449606_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443039_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443039_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443040_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5340440 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5340440 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 101450632 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005340440 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2473562 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2473562 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2505319 | - | - |
mouse-6 v1.1 (Illumina) | scl25940.13_350-S | - | - |
mouse-6 v1.1 (Illumina) | scl27144.1_335-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 101450632 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5340440 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl25940.13_350-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5340440 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101450632 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2473562 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3143483 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2473562 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2505319 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3143483 | - | - |
