Assay Details
Gene Symbol: Abhd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190437.1 | NP_001177366.1 | 5 - 6 | 726 | 82 | |
NM_145215.2 | NP_660250.1 | 5 - 6 | 1203 | 82 | ||
NR_033794.1 | 4 - 5 | 1188 | 82 | |||
GenBank mRNA | AF412033.1 | - | 5 - 6 | 1203 | 82 | |
AK004244.1 | - | 5 - 6 | 737 | 82 | ||
BC038052.1 | - | 4 - 5 | 605 | 82 | ||
BC051486.1 | - | 1 - 2 | 82 | |||
BC056610.1 | - | 5 - 6 | 865 | 82 | ||
BC056624.1 | - | 4 - 5 | 1188 | 82 | ||
BC069865.1 | - | 2 - 3 | 1190 | 82 | ||
BC069866.1 | - | 5 - 6 | 825 | 6478980 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114261 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114263 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114259 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114265 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114262 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114260 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114258 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P142187 | BC016241.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P142187 | BC069865.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P142187 | BC069866.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P142187 | BC038052.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P142187 | BC056610.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967504 | BC069865.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967504 | BC069866.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967504 | BC016241.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967504 | BC038052.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967504 | BC051486.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 463158 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 519696 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179622 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P142187 | BC016241.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P142187 | BC069865.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P142187 | BC069866.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P142187 | BC038052.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P142187 | BC056610.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967504 | BC069865.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967504 | BC069866.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967504 | BC016241.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967504 | BC038052.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967504 | BC051486.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114242 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114272 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114262 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114268 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114271 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114263 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114267 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114275 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114282 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114283 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114280 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114243 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114276 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114258 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114245 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114250 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114255 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114266 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114265 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114251 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114260 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114274 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114240 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114253 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114278 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179622 | - | - |
Whole Mouse Genome G4122A | A_52_P560620 | BC069865.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P560620 | BC069866.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P560620 | BC016241.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P560620 | BC038052.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P560620 | BC051486.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126683 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179622 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04472304 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04472305 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04472291 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04472298 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04472292 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04472300 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04472297 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04472295 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114258 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114263 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114262 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114259 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114260 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114265 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114271 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114243 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114233 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04399898 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04673711 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114236 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04399904 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114234 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04399906 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04970739 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114282 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114255 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04970736 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114251 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04472292 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04972888 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114268 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114253 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114265 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04399895 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114275 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04238060 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114250 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114244 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04472301 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04951826 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04927068 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114221 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114285 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04970740 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114225 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04820294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04970745 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04472291 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114260 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04972886 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114228 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04597698 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04399908 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04970741 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24114258 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10526255 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10526254 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10526252 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10526251 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10526258 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10526259 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10526257 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10526253 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10526260 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10526256 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1428428_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10526250 | - | - |
Mouse Genome 430 2.0 Array | 1428428_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1428428_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68758_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68758_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500014057.mm.1 | TC0500001572.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500014057.mm.1 | NM_001190437.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014057.mm.1 | NM_001190437.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014057.mm.1 | NM_145215.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014057.mm.1 | NM_145215.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014057.mm.1 | NR_033794.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014057.mm.1 | NR_033794.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014055.mm.1 | TC0500001572.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500014055.mm.1 | NM_001190437.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014055.mm.1 | NM_001190437.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014055.mm.1 | NM_145215.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014055.mm.1 | NM_145215.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014055.mm.1 | NR_033794.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014055.mm.1 | NR_033794.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014056.mm.1 | TC0500001572.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500014056.mm.1 | NM_001190437.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014056.mm.1 | NM_001190437.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014056.mm.1 | NM_145215.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014056.mm.1 | NM_145215.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014056.mm.1 | NR_033794.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500014056.mm.1 | NR_033794.1 | Refseq |
Mouse Expression Set 430 | 1428428_PM_at | - | - |
Mouse Expression Set 430 | 1428428_at | - | - |
Murine Genome U74v2 Set | 165420_PM_at | - | - |
Murine Genome U74v2 Set | 165420_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1428428_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1428428_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 102570603 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1240086 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 102570603 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001240086 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2453209 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2472940 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2453209 | - | - |
mouse-6 v1.1 (Illumina) | scl0004098.1_24-S | - | - |
mouse-6 v1.1 (Illumina) | scl27141.4_222-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1240086 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102570603 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl27141.4_222-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102570603 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1240086 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2453209 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2453209 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl27141.4_222-S | - | - |
