Assay Details
Gene Symbol: Lin54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115010.1 | NP_001108482.1 | 8 - 9 | 1835 | 56 | |
NM_172714.4 | NP_766302.2 | 8 - 9 | 1703 | 56 | ||
GenBank mRNA | AK029288.1 | - | 9 - 10 | 1149 | 56 | |
AK030894.1 | - | 8 - 9 | 1837 | 56 | ||
AK134320.1 | - | 8 - 9 | 1701 | 56 | ||
AK220225.1 | - | 8 - 9 | 1698 | 56 | ||
BC106126.1 | - | 7 - 8 | 1339 | 56 | ||
BC125580.1 | - | 6 - 7 | 830 | 56 | ||
BC145025.1 | - | 6 - 7 | 830 | 9053573 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
