Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500025000.mm.1 |
TC0500002836.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500025000.mm.1 |
NM_001115010.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500025000.mm.1 |
NM_001115010.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500025000.mm.1 |
NM_172714.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500025000.mm.1 |
NM_172714.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024999.mm.1 |
TC0500002836.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024999.mm.1 |
NM_001115010.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024999.mm.1 |
NM_001115010.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024999.mm.1 |
NM_172714.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024999.mm.1 |
NM_172714.4 |
Refseq |