Assay Details
Gene Symbol: Fbrsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142642.1 | NP_001136114.1 | 17 - 18 | 2387 | 93 | |
NM_028596.2 | NP_082872.2 | 10 - 11 | 1431 | 93 | ||
GenBank mRNA | AK031967.1 | - | 10 - 11 | 1401 | 93 | |
AK129388.1 | - | 6 - 7 | 2943 | 93 | ||
BC059932.1 | - | 15 - 16 | 3173 | 93 | ||
BC069994.1 | - | 5 - 6 | 388 | 93 | ||
BC151196.1 | - | 12 - 13 | 1396 | 9088084 | 93 | |
BU614458.1 | - | 7 - 8 | 623 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
