Assay Details
Gene Symbol: Fbrsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142642.1 | NP_001136114.1 | 4 - 5 | 962 | 58 | |
GenBank mRNA | BC059932.1 | - | 4 - 5 | 945 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
