Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500026456.mm.1 |
TC0500002989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500026456.mm.1 |
NM_001142642.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500026456.mm.1 |
NM_001142642.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500026457.mm.1 |
TC0500002989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500026457.mm.1 |
NM_001142642.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500026457.mm.1 |
NM_001142642.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500026457.mm.1 |
NM_028596.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500026457.mm.1 |
NM_028596.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500026461.mm.1 |
TC0500002989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500026461.mm.1 |
NM_028596.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500026461.mm.1 |
NM_028596.2 |
Refseq |