Assay Details
| Gene Symbol: Galnt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198306.3 | NP_938048.2 | 1 - 2 | 585 | 63 | |
| GenBank mRNA | AK032568.1 | - | 1 - 2 | 586 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt9
Genomic Map