Assay Details
| Gene Symbol: Tnrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122730.2 | NP_001116202.2 | 1 - 2 | 114 | 108 | |
| NM_178242.2 | NP_839973.1 | 1 - 2 | 114 | 108 | ||
| GenBank mRNA | BC049818.1 | - | 1 - 2 | 114 | 6511384 | 108 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC049818.1, NM_001122730.2, NM_178242.2 transcripts.
Set Membership: |
|

Tnrc18
Genomic Map