Assay Details
| Gene Symbol: Tle4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF229633.1 | - | 18 - 19 | 2149 | 65 | |
| AK122481.2 | - | 16 - 17 | 2008 | 65 | ||
| AK134989.1 | - | 18 - 19 | 2783 | 65 | ||
| BC019750.1 | - | 2 - 3 | 214 | 65 | ||
| BC043477.1 | - | 3 - 4 | 363 | 65 | ||
| BC058525.1 | - | 1 - 2 | 102 | 65 | ||
| BC086631.1 | - | 19 - 20 | 2692 | 65 | ||
| BC086780.1 | - | 1 - 2 | 42 | 65 | ||
| CB058377.1 | - | 1 - 2 | 56 | 65 | ||
| U61363.1 | - | 10 - 11 | 1552 | 65 | ||
More Information
Set Membership: |
|

Tle4
Genomic Map