Assay Details
Gene Symbol: Mvk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF137598.1 | - | 7 - 8 | 788 | 110 | |
AK031084.1 | - | 7 - 8 | 1568 | 110 | ||
AK149630.1 | - | 8 - 9 | 885 | 110 | ||
AK177080.1 | - | 4 - 5 | 270 | 110 | ||
BC005606.1 | - | 8 - 9 | 841 | 3498140 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
