Assay Details
| Gene Symbol: Gda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010266.2 | NP_034396.1 | 2 - 3 | 388 | 79 | |
| GenBank mRNA | AF369880.1 | - | 2 - 3 | 273 | 79 | |
| AF369881.1 | - | 2 - 3 | 273 | 79 | ||
| AF369882.1 | - | 2 - 3 | 273 | 79 | ||
| AF369883.1 | - | 2 - 3 | 273 | 79 | ||
| AF369884.1 | - | 2 - 3 | 273 | 79 | ||
| AK041056.1 | - | 2 - 3 | 327 | 79 | ||
| AK044078.1 | - | 2 - 3 | 316 | 79 | ||
| AK079419.1 | - | 2 - 3 | 196 | 79 | ||
| AK079459.1 | - | 2 - 3 | 328 | 79 | ||
| AK139617.1 | - | 155 | 79 | |||
| AK157014.1 | - | 2 - 3 | 350 | 79 | ||
| AK173138.1 | - | 2 - 3 | 317 | 79 | ||
| BC076583.1 | - | 2 - 3 | 301 | 30620457 | 79 | |
More Information
Set Membership: |
|

Gda
Genomic Map