Assay Details
| Gene Symbol: Gda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010266.2 | NP_034396.1 | 7 - 8 | 888 | 66 | |
| GenBank mRNA | AF174583.1 | - | 7 - 8 | 814 | 66 | |
| AF369880.1 | - | 7 - 8 | 773 | 66 | ||
| AF369881.1 | - | 7 - 8 | 773 | 66 | ||
| AF369882.1 | - | 7 - 8 | 773 | 66 | ||
| AF369883.1 | - | 7 - 8 | 773 | 66 | ||
| AF369884.1 | - | 7 - 8 | 773 | 66 | ||
| AK041056.1 | - | 7 - 8 | 827 | 66 | ||
| AK079419.1 | - | 7 - 8 | 696 | 66 | ||
| AK079459.1 | - | 7 - 8 | 828 | 66 | ||
| AK139617.1 | - | 656 | 66 | |||
| AK157014.1 | - | 7 - 8 | 850 | 66 | ||
| AK173138.1 | - | 7 - 8 | 817 | 66 | ||
| BC076583.1 | - | 7 - 8 | 801 | 30620457 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gda
Genomic Map