Assay Details
| Gene Symbol: Sec31b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033343.1 | NP_001028515.1 | 7 - 8 | 876 | 68 | |
| GenBank mRNA | AK158017.1 | - | 6 - 7 | 753 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sec31b
Genomic Map