Assay Details
| Gene Symbol: Kcna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008417.5 | NP_032443.3 | 1 - 2 | 196 | 93 | |
| GenBank mRNA | AK044342.1 | - | 1 - 2 | 197 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK044342.1, NM_008417.5 transcripts.
Set Membership: |
|

Kcna2
Genomic Map