Assay Details
| Gene Symbol: Pi4k2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145501.2 | NP_663476.1 | 7 - 8 | 1291 | 91 | |
| GenBank mRNA | AK030725.1 | - | 7 - 8 | 1256 | 91 | |
| AK165136.1 | - | 1 - 2 | 1347 | 91 | ||
| BC089309.1 | - | 6 - 7 | 1980 | 91 | ||
| BC110363.1 | - | 7 - 8 | 1253 | 5346917 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pi4k2a
Genomic Map