Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700027018.mm.1 |
NM_201374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027018.mm.1 |
NM_201374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027018.mm.1 |
TC0700002996.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027021.mm.1 |
NM_201374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027021.mm.1 |
NM_201374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027021.mm.1 |
TC0700002996.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027013.mm.1 |
NM_201374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027013.mm.1 |
NM_201374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027013.mm.1 |
TC0700002996.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027014.mm.1 |
NM_201374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027014.mm.1 |
NM_201374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027014.mm.1 |
TC0700002996.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027017.mm.1 |
NM_201374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027017.mm.1 |
NM_201374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027017.mm.1 |
TC0700002996.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027020.mm.1 |
NM_201374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027020.mm.1 |
NM_201374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027020.mm.1 |
TC0700002996.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027022.mm.1 |
NM_201374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027022.mm.1 |
NM_201374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027022.mm.1 |
TC0700002996.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027019.mm.1 |
NM_201374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027019.mm.1 |
NM_201374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027019.mm.1 |
TC0700002996.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027016.mm.1 |
NM_201374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027016.mm.1 |
NM_201374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027016.mm.1 |
TC0700002996.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027023.mm.1 |
NM_201374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027023.mm.1 |
NM_201374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027023.mm.1 |
TC0700002996.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027024.mm.1 |
NM_201374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027024.mm.1 |
NM_201374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027024.mm.1 |
TC0700002996.mm.1 |
MTA TC |