Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700027010.mm.1 |
NM_201374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027010.mm.1 |
NM_201374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027010.mm.1 |
TC0700002996.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027012.mm.1 |
NM_201374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027012.mm.1 |
NM_201374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027012.mm.1 |
TC0700002996.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027009.mm.1 |
NM_201374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027009.mm.1 |
NM_201374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027009.mm.1 |
TC0700002996.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027011.mm.1 |
NM_201374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027011.mm.1 |
NM_201374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027011.mm.1 |
TC0700002996.mm.1 |
MTA TC |