Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500012989.mm.1 |
TC0500001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012989.mm.1 |
NM_001190795.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012989.mm.1 |
NM_001190795.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012989.mm.1 |
NM_026409.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012989.mm.1 |
NM_026409.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012984.mm.1 |
TC0500001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012984.mm.1 |
NM_001190795.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012984.mm.1 |
NM_001190795.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012984.mm.1 |
NM_026409.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012984.mm.1 |
NM_026409.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012987.mm.1 |
TC0500001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012987.mm.1 |
NM_001190795.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012987.mm.1 |
NM_001190795.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012987.mm.1 |
NM_026409.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012987.mm.1 |
NM_026409.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012991.mm.1 |
TC0500001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012991.mm.1 |
NM_001190795.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012991.mm.1 |
NM_001190795.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012991.mm.1 |
NM_026409.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012991.mm.1 |
NM_026409.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012988.mm.1 |
TC0500001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012988.mm.1 |
NM_001190795.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012988.mm.1 |
NM_001190795.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012988.mm.1 |
NM_026409.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012988.mm.1 |
NM_026409.4 |
Refseq |