Assay Details
Gene Symbol: Ddx55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190795.1 | NP_001177724.1 | 6 - 7 | 619 | 87 | |
NM_026409.4 | NP_080685.2 | 6 - 7 | 619 | 87 | ||
GenBank mRNA | AK012782.1 | - | 6 - 7 | 87 | ||
AK017554.1 | - | 6 - 7 | 561 | 87 | ||
AK033741.1 | - | 6 - 7 | 596 | 87 | ||
AK077525.1 | - | 6 - 7 | 600 | 87 | ||
AK129402.1 | - | 6 - 7 | 597 | 87 | ||
AK154422.1 | - | 6 - 7 | 620 | 87 | ||
AK167149.1 | - | 5 - 6 | 2614 | 87 | ||
BC043052.1 | - | 6 - 7 | 569 | 87 | ||
BC117786.1 | - | 6 - 7 | 581 | 40092124 | 87 | |
BC117787.1 | - | 6 - 7 | 581 | 40092125 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
