Assay Details
Gene Symbol: Zfp719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172482.1 | NP_766070.1 | 1 - 2 | 179 | 67 | |
GenBank mRNA | AK044684.1 | - | 1 - 2 | 156 | 67 | |
AK083143.1 | - | 1 - 2 | 120 | 67 | ||
BC081548.1 | - | 1 - 2 | 142 | 67 | ||
CJ086175.1 | - | 1 - 2 | 178 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK083143.1, NM_172482.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
