| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700006202.mm.1 |
TC0700000765.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700006202.mm.1 |
NM_172482.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006202.mm.1 |
NM_172482.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006203.mm.1 |
TC0700000765.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700006203.mm.1 |
NM_172482.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006203.mm.1 |
NM_172482.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006208.mm.1 |
TC0700000765.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700006208.mm.1 |
NM_172482.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006208.mm.1 |
NM_172482.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006212.mm.1 |
TC0700000765.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700006212.mm.1 |
NM_172482.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006212.mm.1 |
NM_172482.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006206.mm.1 |
TC0700000765.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700006206.mm.1 |
NM_172482.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006206.mm.1 |
NM_172482.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006210.mm.1 |
TC0700000765.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700006210.mm.1 |
NM_172482.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006210.mm.1 |
NM_172482.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006209.mm.1 |
TC0700000765.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700006209.mm.1 |
NM_172482.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006209.mm.1 |
NM_172482.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006211.mm.1 |
TC0700000765.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700006211.mm.1 |
NM_172482.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006211.mm.1 |
NM_172482.1 |
Refseq |