Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500013922.mm.1 |
NM_001168288.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013922.mm.1 |
NM_001168288.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013922.mm.1 |
NM_198420.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013922.mm.1 |
NM_198420.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013922.mm.1 |
TC1500001791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500013929.mm.1 |
NM_001168288.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013929.mm.1 |
NM_001168288.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013929.mm.1 |
NM_198420.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013929.mm.1 |
NM_198420.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013929.mm.1 |
TC1500001791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500013935.mm.1 |
NM_001168288.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013935.mm.1 |
NM_001168288.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013935.mm.1 |
NM_198420.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013935.mm.1 |
NM_198420.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013935.mm.1 |
TC1500001791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500013927.mm.1 |
NM_001168288.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013927.mm.1 |
NM_001168288.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013927.mm.1 |
NM_198420.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013927.mm.1 |
NM_198420.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013927.mm.1 |
TC1500001791.mm.1 |
MTA TC |