Assay Details
| Gene Symbol: Arhgap39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168288.1 | NP_001161760.1 | 4 - 5 | 845 | 132 | |
| NM_198420.2 | NP_940812.2 | 4 - 5 | 845 | 132 | ||
| GenBank mRNA | AK035479.1 | - | 3 - 4 | 550 | 132 | |
| AK141623.1 | - | 4 - 5 | 805 | 132 | ||
| AK173232.1 | - | 4 - 5 | 780 | 132 | ||
| BC060637.1 | - | 4 - 5 | 826 | 6849400 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap39
Genomic Map