Assay Details
Gene Symbol: Emc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197991.2 | NP_932108.2 | 2 - 3 | 331 | 69 | |
GenBank mRNA | AK150499.1 | - | 2 - 3 | 236 | 69 | |
AK160274.1 | - | 2 - 3 | 246 | 69 | ||
AK169010.1 | - | 2 - 3 | 282 | 69 | ||
AK171657.1 | - | 2 - 3 | 245 | 69 | ||
AY761096.1 | - | 2 - 3 | 224 | 69 | ||
AY761098.1 | - | 2 - 3 | 224 | 69 | ||
BC020179.1 | - | 2 - 3 | 226 | 4913500 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
