Assay Details
| Gene Symbol: Emc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197991.2 | NP_932108.2 | 5 - 6 | 729 | 106 | |
| GenBank mRNA | AK150499.1 | - | 5 - 6 | 634 | 106 | |
| AK160274.1 | - | 5 - 6 | 646 | 106 | ||
| AK169010.1 | - | 5 - 6 | 680 | 106 | ||
| AK171657.1 | - | 5 - 6 | 106 | |||
| AY761096.1 | - | 5 - 6 | 622 | 106 | ||
| AY761098.1 | - | 5 - 6 | 622 | 106 | ||
| BC006065.1 | - | 3 - 4 | 366 | 106 | ||
| BC020179.1 | - | 5 - 6 | 4913500 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Emc10
Genomic Map