Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900002836.mm.1 |
NM_172635.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002836.mm.1 |
NM_172635.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002836.mm.1 |
TC1900000235.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002833.mm.1 |
NM_172635.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002833.mm.1 |
NM_172635.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002833.mm.1 |
TC1900000235.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002835.mm.1 |
NM_172635.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002835.mm.1 |
NM_172635.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002835.mm.1 |
TC1900000235.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002834.mm.1 |
NM_172635.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002834.mm.1 |
NM_172635.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002834.mm.1 |
TC1900000235.mm.1 |
MTA TC |