Assay Details
Gene Symbol: Sept14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028826.1 | NP_083102.1 | 7 - 8 | 1007 | 67 | |
GenBank mRNA | AK006043.1 | - | 2 - 3 | 198 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
