Assay Details
Gene Symbol: Sept14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028826.1 | NP_083102.1 | 8 - 9 | 1104 | 68 | |
GenBank mRNA | AK006043.1 | - | 3 - 4 | 295 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
