Assay Details
Gene Symbol: Sept14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028826.1 | NP_083102.1 | 9 - 10 | 1277 | 61 | |
GenBank mRNA | AK006043.1 | - | 4 - 5 | 468 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382407 | NM_028826.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382407 | AK006043.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382407 | NM_028826.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382407 | AK006043.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2065606 | NM_028826.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164155 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 161911 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 271242 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 199862 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27495 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 4515 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639288 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 225307 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922245 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 440676 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437372 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8760 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616541 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641639 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596979 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 357932 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92354 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139027 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96446 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119330 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382407 | NM_028826.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382407 | AK006043.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382407 | NM_028826.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382407 | AK006043.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2065606 | NM_028826.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119330 | - | - |
Whole Mouse Genome G4122A | A_51_P382407 | NM_028826.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P382407 | AK006043.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P382407 | NM_028826.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P382407 | AK006043.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090425 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090489 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090472 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04592404 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090457 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090432 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090422 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090468 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04360738 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090487 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090466 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04420392 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04533433 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090437 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090390 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04113992 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090424 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090451 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090475 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04724142 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090423 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090428 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04672088 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04203114 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090385 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090485 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090387 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04592402 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090438 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090453 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04226610 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090395 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090403 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090400 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090486 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090425 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04113997 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090443 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090463 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090439 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04113989 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090440 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090483 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090420 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090398 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04989467 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04448536 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04940240 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090389 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04579110 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090442 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090419 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04465065 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090459 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090472 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090396 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04363482 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090452 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04465064 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090421 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090404 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04724145 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090391 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090464 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090479 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24090454 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10534072 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534070 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534069 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534071 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534067 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534073 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534068 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534074 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1432446_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10534066 | - | - |
Mouse Genome 430 2.0 Array | 1432446_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1432446_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74222_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74222_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500029892.mm.1 | TC0500003306.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500029892.mm.1 | NM_028826.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500029892.mm.1 | NM_028826.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500029891.mm.1 | TC0500003306.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500029891.mm.1 | NM_028826.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500029891.mm.1 | NM_028826.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500029893.mm.1 | TC0500003306.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500029893.mm.1 | NM_028826.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500029893.mm.1 | NM_028826.1 | Refseq |
Mouse Expression Set 430 | 1432446_PM_at | - | - |
Mouse Expression Set 430 | 1432446_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432446_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432446_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 477527 | - | - |
