Assay Details
Gene Symbol: Psph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133900.4 | NP_598661.1 | 2 - 3 | 380 | 74 | |
GenBank mRNA | AK088865.1 | - | 2 - 3 | 332 | 74 | |
CV560928.1 | - | 2 - 3 | 316 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK088865.1, NM_133900.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
