Assay Details
| Gene Symbol: Entpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF037366.1 | - | 4 - 5 | 478 | 93 | |
| AK028625.1 | - | 1 - 2 | 144 | 93 | ||
| AK029512.1 | - | 4 - 5 | 524 | 93 | ||
| AK030601.1 | - | 4 - 5 | 504 | 93 | ||
| BC011278.1 | - | 4 - 5 | 3674983 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Entpd1
Genomic Map