Assay Details
| Gene Symbol: Msr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113326.1 | NP_001106797.1 | 6 - 7 | 971 | 87 | |
| NM_031195.2 | NP_112472.2 | 6 - 7 | 971 | 87 | ||
| GenBank mRNA | AF203781.1 | - | 5 - 6 | 913 | 87 | |
| AK028480.1 | - | 6 - 7 | 965 | 87 | ||
| AK089178.1 | - | 6 - 7 | 965 | 87 | ||
| AK150518.1 | - | 6 - 7 | 972 | 87 | ||
| AK151403.1 | - | 6 - 7 | 945 | 87 | ||
| AK151855.1 | - | 6 - 7 | 972 | 87 | ||
| AK154806.1 | - | 6 - 7 | 948 | 87 | ||
| AK155362.1 | - | 6 - 7 | 967 | 87 | ||
| AK155589.1 | - | 6 - 7 | 965 | 87 | ||
| BC003814.1 | - | 6 - 7 | 942 | 3594142 | 87 | |
| D13382.1 | - | 5 - 6 | 913 | 87 | ||
| L04274.1 | - | 6 - 7 | 918 | 87 | ||
| L04275.1 | - | 6 - 7 | 918 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msr1
Genomic Map