Assay Details
Gene Symbol: Msr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031195.2 | NP_112472.2 | 8 - 9 | 1112 | 77 | |
GenBank mRNA | AK028480.1 | - | 8 - 9 | 1106 | 77 | |
AK089178.1 | - | 8 - 9 | 1106 | 77 | ||
AK150518.1 | - | 8 - 9 | 1113 | 77 | ||
AK151403.1 | - | 8 - 9 | 1086 | 77 | ||
AK151855.1 | - | 8 - 9 | 1113 | 77 | ||
AK155362.1 | - | 8 - 9 | 1108 | 77 | ||
AK155589.1 | - | 8 - 9 | 1106 | 77 | ||
BC003814.1 | - | 8 - 9 | 1083 | 3594142 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
