Assay Details
| Gene Symbol: Tm9sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133352.2 | NP_579930.1 | 8 - 9 | 1197 | 78 | |
| GenBank mRNA | AF269151.1 | - | 8 - 9 | 1129 | 78 | |
| AK147380.1 | - | 8 - 9 | 1192 | 78 | ||
| AK147629.1 | - | 8 - 9 | 1191 | 78 | ||
| AK220279.1 | - | 8 - 9 | 1173 | 78 | ||
| BC004799.1 | - | 7 - 8 | 826 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tm9sf3
Genomic Map