Assay Details
| Gene Symbol: Leprotl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026609.2 | NP_080885.1 | 1 - 2 | 50 | 70 | |
| GenBank mRNA | AK003329.1 | - | 1 - 2 | 61 | 70 | |
| AK013870.1 | - | 1 - 2 | 120 | 70 | ||
| AK075740.1 | - | 1 - 2 | 105 | 70 | ||
| AK150289.1 | - | 1 - 2 | 109 | 70 | ||
| AK153277.1 | - | 1 - 2 | 95 | 70 | ||
| AK154712.1 | - | 1 - 2 | 115 | 70 | ||
| AK159401.1 | - | 1 - 2 | 38 | 70 | ||
| BC004677.1 | - | 1 - 2 | 50 | 3500223 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003329.1, AK013870.1, AK075740.1, AK150289.1, AK153277.1, AK154712.1, AK159401.1, BC004677.1, NM_026609.2 transcripts.
Set Membership: |
|

Leprotl1
Genomic Map